Methyl-ɑ-D-galactopyranoside monohydrate ≥98.0%

Supplier: TCI America

M1047-25G M1047-5G
TCM1047-25GEA 229.1 CAD
TCM1047-25G TCM1047-005G
Methyl-ɑ-D-galactopyranoside monohydrate ≥98.0%
Methyl-alpha-D-galactopyranoside monohydrate

CAS Number: 34004-14-3
MDL Number: MFCD00064085
Molecular Formula: C7H14O6
Molecular Weight: 194.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 108
Specific rotation [a]20/D: 177 deg (C=1, H2O)

Formula: C₇H₁₄O₆.H₂O
Melting Pt: 108 °C
MDL Number: MFCD00064085
CAS Number: 34004-14-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7d6bca153a199dfd8d75691a13ceb9565272562&vid=e7d6bca153a199dfd8d75691a13ceb9565272562&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR