1-Acetyl-1,2,3,4-tetrahydroquinoline ≥98.0%

Supplier: TCI America

Synonyms: 1,2,3,4-Tetrahydro-1-acetylquinoline

A1641-5G A1641-25G
TCA1641-5GEA 115.66 CAD
TCA1641-5G TCA1641-25G
1-Acetyl-1,2,3,4-tetrahydroquinoline ≥98.0%
1-Acetyl-1,2,3,4-tetrahydroquinoline

CAS Number: 4169-19-1
MDL Number: MFCD01195305
Molecular Formula: C11H13NO
Molecular Weight: 175.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 295
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: 1,2,3,4-Tetrahydro-1-acetylquinoline

Formula: C₁₁H₁₃NO
MW: 175.23 g/mol
Boiling Pt: 295 °C
Density: 1.12 (20 °C)
MDL Number: MFCD01195305
CAS Number: 4169-19-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2378e6aa5199fa58f74434ec3d55a74c7cd35ca7&vid=2378e6aa5199fa58f74434ec3d55a74c7cd35ca7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR