Methoxybenzoquinone ≥99.0%

Supplier: TCI America

M1045-1G M1045-5G
TCM1045-001GEA 212.24 CAD
TCM1045-001G TCM1045-005G
Methoxybenzoquinone ≥99.0%
Methoxybenzoquinone

CAS Number: 2880-58-2
MDL Number: MFCD00144755
Molecular Formula: C7H6O3
Molecular Weight: 138.12
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 147

Formula: C₇H₆O₃
Melting Pt: 147 °C
MDL Number: MFCD00144755
CAS Number: 2880-58-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b41b9fcb0af191bb006487308089ff96514841a0&vid=b41b9fcb0af191bb006487308089ff96514841a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR