(S)-(-)-Menthyltoluene-4-sulfinate ≥98.0%

Supplier: TCI America

Synonyms: (S)-((1R,2S,5R)-2-isopropyl-5-methylcyclohexyl) 4-methylbenzenesulfinate, (1R,2S,5R)-(-)-Menthyl (S)-p-toluenesulfinate, (1R,2S,5R)-(-)-Menthyl (S)-p-toluenesulphinate, (S)-((1R,2S,5R)-2-isopropyl-5-methylcyclohexyl) 4-methylbenzenesulphinate

M1044-5G M1044-25G
TCM1044-005GEA 171.9 CAD
TCM1044-005G TCM1044-25G
(S)-(-)-Menthyltoluene-4-sulfinate ≥98.0%
(S)-(-)-Menthyltoluene-4-sulphinate

CAS Number: 1517-82-4
MDL Number: MFCD00010192
Molecular Formula: C17H26O2S
Molecular Weight: 294.45
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 105
Specific rotation [a]20/D: -200 deg (C=2, Acetone)
Storage Temperature: <0°C

Formula: C₁₇H₂₆O₂S
Melting Pt: 105 °C
MDL Number: MFCD00010192
CAS Number: 1517-82-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1858c10b16e9ab82d016ce910e23edde41491d3d&vid=1858c10b16e9ab82d016ce910e23edde41491d3d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR