4-Hydroxy-1-methyl-2-quinolone ≥98.0%
Supplier: TCI America
Synonyms:
4-Hydroxy-1-methyl-2(1H)-quinolone
M1009-25G
M1009-5G
TCM1009-025GEA
147.1
CAD
TCM1009-025G
TCM1009-5G
4-Hydroxy-1-methyl-2-quinolone ≥98.0%
4-Hydroxy-1-methyl-2-quinolone
CAS Number: 1677-46-9
MDL Number: MFCD00024052
Molecular Formula: C10H9NO2
Molecular Weight: 175.19
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 270
Formula:
C₁₀H₉NO₂ Melting Pt: 270 °C |
MDL Number:
MFCD00024052 CAS Number: 1677-46-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...