2-Hydroxy-N-methylbenzylamine hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
N-Methyl-2-hydroxybenzylamine Hydrochloride, 2-(Methylaminomethyl)phenol Hydrochloride, alpha-Methylamino-o-cresol Hydrochloride
CAS Number: 60399-02-2
MDL Number: MFCD00142934
Molecular Formula: C8H12NO
Molecular Weight: 173.64
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 148
Other Product Names / Synonyms: alpha-Methylamino-o-cresol Hydrochloride, 2-(Methylaminomethyl)phenol Hydrochloride, N-Methyl-2-hydroxybenzylamine Hydrochloride
Formula:
C₈H₁₂NO Melting Pt: 148 °C |
MDL Number:
MFCD00142934 CAS Number: 60399-02-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...