4-Methoxy-4'-nitrobiphenyl ≥98.0%

Supplier: TCI America

Synonyms: 4-(4'-Nitrophenyl)anisole, 4-(4'-Methoxyphenyl)nitrobenzene, 4-Methoxy-4'-nitro-1,1'-biphenyl

M0997-1G M0997-100MG
TCM0997-001GEA 290.72 CAD
TCM0997-001G TCM0997-100MG
4-Methoxy-4'-nitrobiphenyl ≥98.0%
4-Methoxy-4'-nitrobiphenyl

CAS Number: 2143-90-0
MDL Number: MFCD00143334
Molecular Formula: C13H11NO3
Molecular Weight: 229.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 110


Product Categories: Materials Chemistry, Organic Nonlinear Optical (NLO) Materials
Other Product Names / Synonyms: 4-(4'-Methoxyphenyl)nitrobenzene, 4-(4'-Nitrophenyl)anisole

Formula: C₁₃H₁₁NO₃
MW: 229.24 g/mol
Melting Pt: 110 °C
MDL Number: MFCD00143334
CAS Number: 2143-90-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16ba10e12dc51edd57d0345e26d3fea839ac4833&vid=16ba10e12dc51edd57d0345e26d3fea839ac4833&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR