Methyl (methoxymethylene)acetate ≥95.0%
Supplier: TCI America
M0995-500ML
M0995-25ML
TCM0995-500MLEA
325.48
CAD
TCM0995-500ML
TCM0995-025ML
Methyl (methoxymethylene)acetate ≥95.0%
Methyl (methoxymethylene)acetate
CAS Number: 34846-90-7
MDL Number: MFCD00128206
Molecular Formula: C5H8O3
Molecular Weight: 116.12
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 56
Flash Point (°C): 65
Specific Gravity (20/20): 1.07
Other Product Names / Synonyms: 3-Methoxyacrylic Acid Methyl Ester
Formula:
C₅H₈O₃ Boiling Pt: 56 °C Density: 1.07 (20 °C) Flash Pt: 65 °C |
MDL Number:
MFCD00128206 CAS Number: 34846-90-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...