N-Methyl-1-naphthylmethylamine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1-(Methylaminomethyl)naphthalene

M0983-5G M0983-1G
TCM0983-005GEA 164.32 CAD
TCM0983-005G TCM0983-001G
N-Methyl-1-naphthylmethylamine ≥98.0% (by titrimetric analysis)
N-Methyl-1-naphthylmethylamine

CAS Number: 14489-75-9
MDL Number: MFCD00144934
Molecular Formula: C12H13N
Molecular Weight: 171.24
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 120
Specific Gravity (20/20): 1.05


Other Product Names / Synonyms: 1-(Methylaminomethyl)naphthalene

Formula: C₁₂H₁₃N
MW: 171.24 g/mol
Boiling Pt: 120 °C
Density: 1.05 (20 °C)
MDL Number: MFCD00144934
CAS Number: 14489-75-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=daa05759d85508f9a4e9aa0822e9f5b89b2de875&vid=daa05759d85508f9a4e9aa0822e9f5b89b2de875&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR