Potassium ethyl malonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

M0975-25G M0975-250G
TCM0975-025GEA 88.43 CAD
TCM0975-025G TCM0975-250G
Potassium ethyl malonate ≥98.0% (by titrimetric analysis)
Potassium ethyl malonate

CAS Number: 6148-64-7
MDL Number: MFCD00035603
Molecular Formula: C5H8O4
Molecular Weight: 170.21
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 200


Other Product Names / Synonyms: Malonic Acid Monoethyl Ester Potassium Salt, Ethyl Potassium Malonate, Malonic Acid Ethyl Ester Potassium Salt, Monoethyl Malonate Potassium Salt

Formula: C₅H₇KO₄
Melting Pt: 200 °C
MDL Number: MFCD00035603
CAS Number: 6148-64-7
EINECS: 228-156-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6485bc064da43ec100c7a38b2fd8408a383489f7&vid=6485bc064da43ec100c7a38b2fd8408a383489f7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR