4-Methoxyphenylacetic anhydride ≥98.0%

Supplier: TCI America

M0968-500G M0968-25G
TCM0968-500GEA 1110.74 CAD
TCM0968-500G TCM0968-025G
4-Methoxyphenylacetic anhydride ≥98.0%
4-Methoxyphenylacetic anhydride

CAS Number: 3951-10-8
MDL Number: MFCD00059089
Molecular Formula: C18H18O5
Molecular Weight: 314.34
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 76

Formula: C₁₈H₁₈O₅
Melting Pt: 76 °C
MDL Number: MFCD00059089
CAS Number: 3951-10-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=996df776407663709ccd9e7c5e6e7230faa05476&vid=996df776407663709ccd9e7c5e6e7230faa05476&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR