(S)-(+)-6-Methyl-1-octanol ≥98.0%

Supplier: TCI America

M0966-1ML M0966-5ML
TCM0966-001MLEA 1444.59 CAD
TCM0966-001ML TCM0966-005ML
(S)-(+)-6-Methyl-1-octanol ≥98.0%
(S)-(+)-6-Methyl-1-octanol

CAS Number: 110453-78-6
MDL Number: MFCD00221493
Molecular Formula: C9H20O
Molecular Weight: 144.26
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 208
Specific Gravity (20/20): 0.83

Formula: C₉H₂₀O
Boiling Pt: 208 °C
Density: 0,83 (20 °C)
MDL Number: MFCD00221493
CAS Number: 110453-78-6

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR