(S)-(+)-4-Methyl-1-hexanol ≥98.0%

Supplier: TCI America

M0964-1ML M0964-5ML
TCM0964-001MLEA 249.64 CAD
TCM0964-001ML TCM0964-005ML
(S)-(+)-4-Methyl-1-hexanol ≥98.0%
(S)-(+)-4-Methyl-1-hexanol

CAS Number: 1767-46-0
MDL Number: MFCD00221492
Molecular Formula: C7H16O
Molecular Weight: 116.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 126
Specific Gravity (20/20): 0.83
Specific rotation [a]20/D: 7.5 deg (neat)

Formula: C₇H₁₆O
Boiling Pt: 126 °C
Density: 0,83 (20 °C)
MDL Number: MFCD00221492
CAS Number: 1767-46-0

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR