(1S,2R)-1-Amino-2,3-dihydro-1H-inden-2-ol ≥98.0%
Supplier: TCI America
Synonyms:
(1S,2R)-(-)-cis-1-Amino-2-indanol, (1S,2R)-(-)-cis-1-Aminoindan-2-ol
A1624-1G
A1624-5G
TCA1624-1GEA
48.98
CAD
TCA1624-1G
TCA1624-5G
(1S,2R)-1-Amino-2,3-dihydro-1H-inden-2-ol ≥98.0%
(1S,2R)-1-Amino-2,3-dihydro-1H-inden-2-ol
CAS Number: 126456-43-7
MDL Number: MFCD00216655
Molecular Formula: C9H11NO
Molecular Weight: 149.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 117
Specific rotation [a]20/D: -43 deg (C=1, MeOH)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (1S,2R)-(-)-1-Amino-2-hydroxyindan
Formula:
C₉H₁₁NO Melting Pt: 117 °C |
MDL Number:
MFCD00216655 CAS Number: 126456-43-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...