9-Methylcarbazole ≥99.0%

Supplier: TCI America

Synonyms: 9-Methyl-9H-carbazole, N-Methylcarbazole

M0960-5G M0960-25G
TCM0960-005GEA 112.18 CAD
TCM0960-005G TCM0960-025G
9-Methylcarbazole ≥99.0%
9-Methylcarbazole

CAS Number: 1484-12-4
MDL Number: MFCD00013431
Molecular Formula: C13H11N
Molecular Weight: 181.24
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 195
Melting point (°C): 89

Formula: C₁₃H₁₁N
Boiling Pt: 195 °C
Melting Pt: 89 °C
MDL Number: MFCD00013431
CAS Number: 1484-12-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fffe93bd96daa79faf31ffdd3db7ae92edbf48df&vid=fffe93bd96daa79faf31ffdd3db7ae92edbf48df&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR