(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (1R,2S)-(+)-cis-1-Amino-2-indanol, (1R,2S)-(+)-cis-1-Aminoindan-2-ol

A1623-1G A1623-5G
TCA1623-1GEA 45.82 CAD
TCA1623-1G TCA1623-5G
(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol ≥98.0% (by GC, titration analysis)
(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol

CAS Number: 136030-00-7
Molecular Formula: C9H11NO
Molecular Weight: 149.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 116
Specific rotation [a]20/D: 43 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (1R,2S)-(+)-1-Amino-2-hydroxyindan

Formula: C₉H₁₁NO
MW: 149.19 g/mol
Melting Pt: 116 °C
MDL Number: MFCD00216656
CAS Number: 136030-00-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1b3bafa9f943726531c0b4a65f88a09777079e0&vid=c1b3bafa9f943726531c0b4a65f88a09777079e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR