2-Methyl-5-nitroimidazole-1-ethanol ≥99.0% (by GC, titration analysis)

Supplier: TCI America

M0924-25G M0924-500G
TCM0924-025GEA 172.14 CAD
TCM0924-025G TCM0924-500G
2-Methyl-5-nitroimidazole-1-ethanol ≥99.0% (by GC, titration analysis)
2-Methyl-5-nitroimidazole-1-ethanol

CAS Number: 443-48-1
MDL Number: MFCD00009750
Molecular Formula: C6H9N3O3
Molecular Weight: 171.16
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 161


Product Categories: Bioscience, Antibiotic Ingredients, Antiprotozoal Ingredients, Anthelmintic Ingredients
Other Product Names / Synonyms: Metronidazole

Formula: C₆H₉N₃O₃
Melting Pt: 161 °C
MDL Number: MFCD00009750
CAS Number: 443-48-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a05eb09fb5de3f1943dd1b692a28c61733b14656&vid=a05eb09fb5de3f1943dd1b692a28c61733b14656&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR