N-Methyloctadecylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M0912-25G M0912-5G
TCM0912-025GEA 282.82 CAD
TCM0912-025G TCM0912-5G
N-Methyloctadecylamine ≥98.0% (by GC, titration analysis)
N-Methyloctadecylamine

CAS Number: 2439-55-6
MDL Number: MFCD00009008
Molecular Formula: C19H41N
Molecular Weight: 283.54
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 44
Flash Point (°C): 113


Other Product Names / Synonyms: N-Methylstearylamine

Formula: C₁₉H₄₁N
Melting Pt: 44 °C
Flash Pt: 113 °C
MDL Number: MFCD00009008
CAS Number: 2439-55-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8c4493675087c6a2115727e30ea3c62ebc8992d&vid=c8c4493675087c6a2115727e30ea3c62ebc8992d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR