ɑ-Methylene-γ-butyrolactone ≥95.0% stabilized

Supplier: TCI America

M0907-1G M0907-25G M0907-5G
TCM0907-001GEA 91.64 CAD
TCM0907-001G TCM0907-25G TCM0907-5G
ɑ-Methylene-γ-butyrolactone ≥95.0% stabilized
alpha-Methylene-gamma-butyrolactone

(stabilized with 2,6-Di-tert-butyl-p-cresol)
CAS Number: 547-65-9
MDL Number: MFCD00005407
Molecular Formula: C5H6O2
Molecular Weight: 98.10
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): 90
Specific Gravity (20/20): 1.12
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 3-Methylenedihydro-2(3H)-furanone

Formula: C₅H₆O₂
Boiling Pt: 90 °C
Density: 1.12 (20 °C)
Flash Pt: 90 °C
MDL Number: MFCD00005407
CAS Number: 547-65-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c05a1471bd7290520898bd0d1cfb58ba675f78c&vid=1c05a1471bd7290520898bd0d1cfb58ba675f78c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR