DL-O-Methylserine 98,0

Supplier: TCI America

Synonyms: 2-Amino-3-methoxypropanoic acid

M0888-5G M0888-1G
TCM0888-5GEA 856.36 CAD
TCM0888-5G TCM0888-001G
DL-O-Methylserine 98,0
DL-O-Methylserine

CAS Number: 19794-53-7
MDL Number: MFCD00037773
Molecular Formula: C4H9NO3
Molecular Weight: 119.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₄H₉NO₃
MW: 119.12 g/mol
MDL Number: MFCD00037773
CAS Number: 19794-53-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8ed08e0c63989238f7d953ac3096cd9dc57cd1a&vid=e8ed08e0c63989238f7d953ac3096cd9dc57cd1a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR