2-Methyl-5-isopropylaniline ≥93.0%
Supplier: TCI America
M0849-5G
M0849-1G
TCM0849-5GEA
950.37
CAD
TCM0849-5G
TCM0849-001G
2-Methyl-5-isopropylaniline ≥93.0%
2-Methyl-5-isopropylaniline
CAS Number: 2051-53-8
MDL Number: MFCD00044503
Molecular Formula: C10H15N
Molecular Weight: 149.24
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.95
Other Product Names / Synonyms: 2-Amino-p-cymene, 5-Isopropyl-2-methylaniline, 5-Isopropyl-o-toluidine
Formula:
C₁₀H₁₅N Density: 0.95 (20 °C) |
MDL Number:
MFCD00044503 CAS Number: 2051-53-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...