5-Mercapto-1H-1,2,3-triazole sodium salt ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
1H-1,2,3-Triazole-5-thiol Sodium Salt, Sodium 1,2,3-triazole-5-thiolate
M0847-25G
M0847-5G
TCM0847-25GEA
406.06
CAD
TCM0847-25G
TCM0847-5G
5-Mercapto-1H-1,2,3-triazole sodium salt ≥97.0% (by titrimetric analysis)
5-Mercapto-1H-1,2,3-triazole sodium salt
CAS Number: 59032-27-8
MDL Number: MFCD00044799
Molecular Formula: C2H3N3S
Molecular Weight: 123.11
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Other Product Names / Synonyms: 1H-1,2,3-Triazole-5-thiol Sodium Salt
Formula:
C₂H₃N₃S MW: 123.11 g/mol |
MDL Number:
MFCD00044799 CAS Number: 59032-27-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...