Methyl-1-methylpyrrole-2-acetate ≥97.0%

Supplier: TCI America

Synonyms: Methyl 2-(1-methyl-1H-pyrrol-2-yl)acetate, 1-Methylpyrrole-2-acetic Acid Methyl Ester

M0846-25ML M0846-5ML
TCM0846-025MLEA 213.3 CAD
TCM0846-025ML TCM0846-5ML
Methyl-1-methylpyrrole-2-acetate ≥97.0%
Methyl-1-methylpyrrole-2-acetate

CAS Number: 51856-79-2
MDL Number: MFCD00009752
Molecular Formula: C8H11NO2
Molecular Weight: 153.18
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 85
Flash Point (°C): 106
Specific Gravity (20/20): 1.09


Other Product Names / Synonyms: 1-Methylpyrrole-2-acetic Acid Methyl Ester

Formula: C₈H₁₁NO₂
Boiling Pt: 85 °C
Density: 1.09 (20 °C)
Flash Pt: 106 °C
MDL Number: MFCD00009752
CAS Number: 51856-79-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=852b846165958b336769bb3a1e0468bc93fd173e&vid=852b846165958b336769bb3a1e0468bc93fd173e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR