(±)-α-Methoxyphenylacetic acid ≥98.0% for synthesis

Supplier: TCI America

Synonyms: DL-α-Methoxyphenylacetic acid, 2-Methoxy-2-phenylacetic acid, α-Methoxyphenylacetic acid

M0837-25G M0837-5G
TCM0837-025GEA 658.86 CAD
TCM0837-025G TCM0837-005G
(±)-α-Methoxyphenylacetic acid ≥98.0% for synthesis
(±)-α-Methoxyphenylacetic acid

CAS Number: 7021-09-2
MDL Number: MFCD00064217
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 137
Melting point (°C): 70


Other Product Names / Synonyms: DL-O-Methylmandelic Acid

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 165
Melting Pt: 70 °C
MDL Number: MFCD00064217
CAS Number: 7021-09-2
EINECS: 230-300-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8e5f9384b331e66753823e0b786121f159fe78d&vid=e8e5f9384b331e66753823e0b786121f159fe78d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR