2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranosyl azide ≥98.0% (by HPLC)
Supplier: TCI America
A1616-5G
A1616-1G
TCA1616-5GEA
510.34
CAD
TCA1616-5G
TCA1616-1G
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranosyl azide ≥98.0% (by HPLC)
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-beta-D-glucopyranosyl azide
CAS Number: 6205-69-2
MDL Number: MFCD00216968
Molecular Formula: C14H20N4O8
Molecular Weight: 372.33
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 167
Storage Temperature: <0°C
Formula:
C₁₄H₂₀N₄O₈ Melting Pt: 167 °C |
MDL Number:
MFCD00216968 CAS Number: 6205-69-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...