Methyl-5-methoxy-3-oxovalerate ≥96.0%
Supplier: TCI America
M0812-5G
M0812-1G
TCM0812-005GEA
1349.32
CAD
TCM0812-005G
TCM0812-001G
Methyl-5-methoxy-3-oxovalerate ≥96.0%
Methyl-5-methoxy-3-oxovalerate
CAS Number: 62462-05-9
MDL Number: MFCD00042840
Molecular Formula: C7H12O4
Molecular Weight: 160.17
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 69
Flash Point (°C): 104
Specific Gravity (20/20): 1.10
Other Product Names / Synonyms: 3-Keto-5-methoxyvaleric Acid Methyl Ester, 5-Methoxy-3-oxovaleric Acid Methyl Ester, Methyl 3-Keto-5-methoxyvalerate
Formula:
C₇H₁₂O₄ Boiling Pt: 69 °C Density: 1.10 (20 °C) Flash Pt: 104 °C |
MDL Number:
MFCD00042840 CAS Number: 62462-05-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...