2-Ethoxy-3-methylpyrazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Ethoxy-2-methylpyrazine

M0809-25G M0809-5G
TCM0809-025GEA 338.12 CAD
TCM0809-025G TCM0809-005G
2-Ethoxy-3-methylpyrazine ≥98.0% (by GC, titration analysis)
2-Ethoxy-3-methylpyrazine

CAS Number: 32737-14-7
MDL Number: MFCD00038025
Molecular Formula: C7H10N2O
Molecular Weight: 138.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 88
Flash Point (°C): 66
Specific Gravity (20/20): 1.04

Formula: C₇H₁₀N₂O
MW: 138.167 g/mol
Boiling Pt: 88 °C
Density: 1.04 (20 °C)
Flash Pt: 66 °C
MDL Number: MFCD00038025
CAS Number: 32737-14-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3642b06ed12c32192cefbb20620583f74115ad94&vid=3642b06ed12c32192cefbb20620583f74115ad94&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR