2-Methyl-3-isopropoxypyrazine ≥97.0%
Supplier: TCI America
Synonyms:
2-Isopropoxy-3-methylpyrazine
M0808-1G
M0808-5G
TCM0808-001GEA
60.04
CAD
TCM0808-001G
TCM0808-5G
2-Methyl-3-isopropoxypyrazine ≥97.0%
2-Methyl-3-isopropoxypyrazine
CAS Number: 94089-22-2
MDL Number: MFCD00059772
Molecular Formula: C8H12N2O
Molecular Weight: 152.20
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.00
Other Product Names / Synonyms: 2-Isopropoxy-3-methylpyrazine
Formula:
C₈H₁₂N₂O Density: 1.00 (20 °C) |
MDL Number:
MFCD00059772 CAS Number: 94089-22-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...