10-Methyl-9(10H)-acridone ≥98.0%
Supplier: TCI America
Synonyms:
N-Methyl-9-acridone, 10-Methylacridin-9(10H)-one
M0792-25G
M0792-5G
TCM0792-025GEA
914.82
CAD
TCM0792-025G
TCM0792-005G
10-Methyl-9(10H)-acridone ≥98.0%
10-Methyl-9(10H)-acridone
CAS Number: 719-54-0
MDL Number: MFCD00005024
Molecular Formula: C14H11NO
Molecular Weight: 209.25
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 203
Other Product Names / Synonyms: N-Methyl-9-acridone
Formula:
C₁₄H₁₁NO Melting Pt: 203 °C |
MDL Number:
MFCD00005024 CAS Number: 719-54-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...