γ-Methyl-γ-decanolactone ≥98.0%

Supplier: TCI America

M0786-500ML M0786-25ML
TCM0786-500MLEA 619.36 CAD
TCM0786-500ML TCM0786-025ML
γ-Methyl-γ-decanolactone ≥98.0%
gamma-Methyl-gamma-decanolactone

CAS Number: 7011-83-8
MDL Number: MFCD00036642
Molecular Formula: C11H20O2
Molecular Weight: 184.28
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 160
Flash Point (°C): 110
Specific Gravity (20/20): 0.95


Other Product Names / Synonyms: gamma-Methyl-gamma-decalactone

Formula: C₁₁H₂₀O₂
Boiling Pt: 160 °C
Density: 0.95 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00036642
CAS Number: 7011-83-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81c039e6ae037cceb4e81d732ae0fd720b1ee5a3&vid=81c039e6ae037cceb4e81d732ae0fd720b1ee5a3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR