6-Methoxy-1,2,3,4-tetrahydronaphthalene ≥98.0%
Supplier: TCI America
M0765-5ML
M0765-25ML
TCM0765-005MLEA
77.42
CAD
TCM0765-005ML
TCM0765-025ML
6-Methoxy-1,2,3,4-tetrahydronaphthalene ≥98.0%
6-Methoxy-1,2,3,4-tetrahydronaphthalene
CAS Number: 1730-48-9
MDL Number: MFCD00001737
Molecular Formula: C11H14O
Molecular Weight: 162.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 131
Flash Point (°C): 112
Specific Gravity (20/20): 1.03
Formula:
C₁₁H₁₄O MW: 162.23 g/mol Boiling Pt: 131 °C Density: 1.03 (20 °C) Flash Pt: 112 °C |
MDL Number:
MFCD00001737 CAS Number: 1730-48-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...