cis-2-Methoxycinnamic acid ≥98.0%

Supplier: TCI America

Synonyms: cis-O-Methyl-o-cumaric Acid, cis-O-Methyl-o-coumaric Acid

M0761-5G M0761-25G
TCM0761-5GEA 170.64 CAD
TCM0761-5G TCM0761-025G
cis-2-Methoxycinnamic acid ≥98.0%
cis-2-Methoxycinnamic acid

CAS Number: 14737-91-8
MDL Number: MFCD00004375
Molecular Formula: C10H10O3
Molecular Weight: 178.19
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 94


Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: cis-O-Methyl-o-coumaric Acid, cis-O-Methyl-o-cumaric Acid

Formula: C₁₀H₁₀O₃
MW: 178.19 g/mol
Melting Pt: 94 °C
MDL Number: MFCD00004375
CAS Number: 14737-91-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4023b86f00c465da296937f6dd9e709fba420eb2&vid=4023b86f00c465da296937f6dd9e709fba420eb2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR