2-Methoxy-2,4-diphenyl-3(2H)-furanone ≥98.0% (by HPLC)
Supplier: TCI America
M0722-1G
M0722-100MG
TCM0722-001GEA
1466.24
CAD
TCM0722-001G
TCM0722-100MG
2-Methoxy-2,4-diphenyl-3(2H)-furanone ≥98.0% (by HPLC)
2-Methoxy-2,4-diphenyl-3(2H)-furanone
CAS Number: 50632-57-0
MDL Number: MFCD00036834
Molecular Formula: C17H14O3
Molecular Weight: 266.30
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 96
Formula:
C₁₇H₁₄O₃ MW: 266.30 g/mol Melting Pt: 96 °C |
MDL Number:
MFCD00036834 CAS Number: 50632-57-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...