N-Methyl-D-glucamine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
M0713-5G
M0713-25G
TCM0713-5GEA
116.92
CAD
TCM0713-5G
TCM0713-025G
N-Methyl-D-glucamine hydrochloride ≥98.0% (by HPLC, titration analysis)
N-Methyl-D-glucamine hydrochloride
[for Buffer]
CAS Number: 35564-86-4
MDL Number: MFCD00060164
Molecular Formula: C7H17NO5
Molecular Weight: 231.67
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 150
Specific rotation [a]20/D: -20.5 deg (C=10, H2O)
Formula:
C₇H₁₇NO₅ Melting Pt: 150 °C |
MDL Number:
MFCD00060164 CAS Number: 35564-86-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...