Benzyl-DL-mandelate ≥98.0%

Supplier: TCI America

M0711-500G M0711-25G
TCM0711-500GEA 476.05 CAD
TCM0711-500G TCM0711-025G
Benzyl-DL-mandelate ≥98.0%
Benzyl-DL-mandelate

CAS Number: 890-98-2
MDL Number: MFCD00021856
Molecular Formula: C15H14O3
Molecular Weight: 242.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 95


Other Product Names / Synonyms: DL-Mandelic Acid Benzyl Ester

Formula: C₁₅H₁₄O₃
Melting Pt: 95 °C
MDL Number: MFCD00021856
CAS Number: 890-98-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22ddb26f024b4f3e2e6bb5511dcf2cbde349d56a&vid=22ddb26f024b4f3e2e6bb5511dcf2cbde349d56a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR