Methyl-β-D-glucopyranoside hemihydrate ≥98.0%

Supplier: TCI America

M0709-25G M0709-5G
TCM0709-025GEA 704.68 CAD
TCM0709-025G TCM0709-005G
Methyl-β-D-glucopyranoside hemihydrate ≥98.0%
Methyl β-D-glucopyranoside hemihydrate

CAS Number: 709-50-2
MDL Number: MFCD00006602
Molecular Formula: C7H14O6
Molecular Weight: 194.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 111
Specific rotation [a]20/D: -33 deg (C=5, H2O)


Product Categories: Bioscience, Glucose, Glycosides
Other Product Names / Synonyms: Methyl beta-D-Glucoside

Formula: C₇H₁₄O₆·0.5H₂O
Melting Pt: 111 °C
MDL Number: MFCD00149442
CAS Number: 7000-27-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a4031a6f292cb1b9756460fef6968babfb0caf3&vid=4a4031a6f292cb1b9756460fef6968babfb0caf3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR