Methyl-1H-benzotriazole (mixture) ≥98.0%

Supplier: TCI America

M0697-25G M0697-500G M0697-100G
TCM0697-025GEA 83.33 CAD
TCM0697-025G TCM0697-500G TCM0697-100G
Methyl-1H-benzotriazole (mixture) ≥98.0%
Methyl-1H-benzotriazole (mixture)

CAS Number: 29385-43-1
MDL Number: MFCD00167158
Molecular Formula: C7H7N3
Molecular Weight: 133.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 87
Storage Temperature: 0-10°C

Formula: C₇H₇N₃
Melting Pt: 87 °C
MDL Number: MFCD00167158
CAS Number: 29385-43-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ec93890ae5b7696b37576755147413baa174e2e&vid=0ec93890ae5b7696b37576755147413baa174e2e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR