Aminoferrocene ≥96.0%

Supplier: TCI America

Synonyms: Ferrocenylamine

A1601-100MG A1601-1G
TCA1601-100MGEA 159.58 CAD
TCA1601-100MG TCA1601-1G
Aminoferrocene ≥96.0%
Aminoferrocene

CAS Number: 1273-82-1
MDL Number: MFCD03844760
Molecular Formula: C10H11FeN
Molecular Weight: 201.05
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Ferrocenylamine

Formula: C₁₀H₂FeN
MW: 191.97 g/mol
MDL Number: MFCD03844760
CAS Number: 1273-82-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ca23c9ce121782affcd621db7fd8c64e5612798&vid=8ca23c9ce121782affcd621db7fd8c64e5612798&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR