Diisopropyl malonate ≥99.0%

Supplier: TCI America

Synonyms: Malonic Acid Diisopropyl Ester

M0648-25ML M0648-500ML
TCM0648-025MLEA 47.4 CAD
TCM0648-025ML TCM0648-500ML
Diisopropyl malonate ≥99.0%
Diisopropyl malonate

CAS Number: 13195-64-7
MDL Number: MFCD00059359
Molecular Formula: C9H16O4
Molecular Weight: 188.22
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 106
Melting point (°C): -51
Flash Point (°C): 95
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: Malonic Acid Diisopropyl Ester

Formula: C₉H₁₆O₄
Boiling Pt: 106 °C
Melting Pt: –51 °C
Density: 0.99 (20 °C)
Flash Pt: 95 °C
MDL Number: MFCD00059359
CAS Number: 13195-64-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d39625e1f2351df52c63e544437129b5e58400b4&vid=d39625e1f2351df52c63e544437129b5e58400b4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR