4-Methyl-2-pentanone oxime

Supplier: TCI America

Synonyms: Isopropylacetone Oxime, Isobutyl Methyl Ketoxime

M0642-500ML M0642-25ML
TCM0642-500MLEA 1242.99 CAD
TCM0642-500ML TCM0642-025ML
4-Methyl-2-pentanone oxime
4-Methyl-2-pentanone oxime

CAS Number: 105-44-2
MDL Number: MFCD00013933
Molecular Formula: C6H13NO
Molecular Weight: 115.18
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 70
Flash Point (°C): 78
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: Isobutyl Methyl Ketoxime, Isopropylacetone Oxime

Formula: C₆H₁₃NO
Boiling Pt: 70 °C
Density: 0.89 (20 °C)
Flash Pt: 78 °C
MDL Number: MFCD00013933
CAS Number: 105-44-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a58eb161f27f4fdb1ff5d739d7600967b3862d92&vid=a58eb161f27f4fdb1ff5d739d7600967b3862d92&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR