4-Pyridinethiol ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Pyridine-4-thiol, 4-Mercaptopyridine

M0630-5G M0630-25G
TCM0630-005GEA 137.37 CAD
TCM0630-005G TCM0630-025G
4-Pyridinethiol ≥97.0% (by titrimetric analysis)
4-Pyridinethiol

CAS Number: 4556-23-4
MDL Number: MFCD00006422
Molecular Formula: C5H5NS
Molecular Weight: 111.16
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 180
Storage Temperature: <0°C


Other Product Names / Synonyms: 4-Pyridinethiol, 4-Pyridyl Mercaptan

Formula: C₅H₅NS
MW: 111.17 g/mol
Melting Pt: 180 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006422
CAS Number: 4556-23-4
EINECS: 224-926-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5b2329877a733392f35926f50bf3c234fff7ae1&vid=d5b2329877a733392f35926f50bf3c234fff7ae1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR