Monomethyl maleate ≥90.0%

Supplier: TCI America

Synonyms: Methyl Hydrogen Maleate, (Z)-4-Methoxy-4-oxobut-2-enoic acid, Maleic Acid Monomethyl Ester

M0624-25ML M0624-500ML M0624-100ML
TCM0624-025MLEA 139.04 CAD
TCM0624-025ML TCM0624-500ML TCM0624-100ML
Monomethyl maleate ≥90.0%
Monomethyl maleate

CAS Number: 3052-50-4
MDL Number: MFCD00045905
Molecular Formula: C5H6O4
Molecular Weight: 130.10
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.26
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Copolymerization Monomers
Other Product Names / Synonyms: Maleic Acid Monomethyl Ester, Methyl Hydrogen Maleate

Formula: C₅H₆O₄
Density: 1.26 (20 °C)
MDL Number: MFCD00045905
CAS Number: 3052-50-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d25baa348ca09251af4504f82e346e3e5b79d263&vid=d25baa348ca09251af4504f82e346e3e5b79d263&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR