Azobenzene-3,3'-dicarboxylic acid ≥95.0%
Supplier: TCI America
Synonyms:
Azobenzene-3,3'-dicarboxylic Acid, 3,3'-Azodibenzoic Acid, 3,3'-Azodibenzoic acid, 3,3'-(Diazene-1,2-diyl)dibenzoic acid
A1598-1G
A1598-5G
TCA1598-1GEA
232.26
CAD
TCA1598-1G
TCA1598-5G
Azobenzene-3,3'-dicarboxylic acid ≥95.0%
Azobenzene-3,3'-dicarboxylic acid
CAS Number: 621-18-1
MDL Number: MFCD03844759
Molecular Formula: C14H10N2O4
Molecular Weight: 270.24
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 3,3'-Azodibenzoic Acid
Formula:
C₁₄H₁₀N₂O₄ MW: 270.24 g/mol |
MDL Number:
MFCD03844759 CAS Number: 621-18-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...