β-(N-Morpholino)ethanesulphonic acid (MES) ≥99.0% for synthesis

Supplier: TCI America

Synonyms: 2-(N-Morpholino)ethanesulphonic acid, 2-(N-Morpholino)ethanesulfonic acid, 2-Morpholinoethanesulfonic acid, 2-Morpholinoethanesulphonic acid

M0606-25G M0606-250G
TCM0606-025GEA 70.86 CAD
TCM0606-025G TCM0606-250G
β-(N-Morpholino)ethanesulphonic acid (MES) ≥99.0% for synthesis
β-(N-Morpholino)ethanesulphonic acid (MES)

CAS Number: 4432-31-9
MDL Number: MFCD00006181
Molecular Formula: C6H13NO4S
Molecular Weight: 195.23
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 316


Product Categories: Bioscience, Good's Buffering Agents
Other Product Names / Synonyms: MES

Formula: C₆H₁₃NO₄S
Melting Pt: 316 °C
Storage Temperature: Ambient
MDL Number: MFCD00006181
CAS Number: 4432-31-9
EINECS: 224-632-3
Merck Index: 13,05929
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2814d50ce8dbab16b641b2ac981f96ec142c9da0&vid=2814d50ce8dbab16b641b2ac981f96ec142c9da0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR