Azobenzene-4,4'-dicarboxylic acid ≥95.0% (by HPLC, titration analysis)
Supplier: TCI America
A1596-1G
A1596-5G
TCA1596-1GEA
239.69
CAD
TCA1596-1G
TCA1596-5G
Azobenzene-4,4'-dicarboxylic acid ≥95.0% (by HPLC, titration analysis)
Azobenzene-4,4'-dicarboxylic acid
CAS Number: 586-91-4
MDL Number: MFCD00496621
Molecular Formula: C14H10N2O4
Molecular Weight: 270.24
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 4,4'-Azodibenzoic Acid
Formula:
C₁₄H₁₀N₂O₄ |
MDL Number:
MFCD00496621 CAS Number: 586-91-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...