Octadecyl methacrylate ≥97.0% stabilized

Supplier: TCI America

M0593-25G M0593-400G
TCM0593-025GEA 42.66 CAD
TCM0593-025G TCM0593-400G
Octadecyl methacrylate ≥97.0% stabilized
Octadecyl methacrylate

(stabilized with MEHQ)
CAS Number: 32360-05-7
MDL Number: MFCD00026683
Molecular Formula: C22H42O2
Molecular Weight: 338.58
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 235
Flash Point (°C): 191
Freezing point (°C): 20
Specific Gravity (20/20): 0.86


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Methacrylic Acid Octadecyl Ester, Octadecyl Methacrylate, Methacrylic Acid Stearyl Ester

Formula: C₂₂H₄₂O₂
Boiling Pt: 235 °C
Density: 0.86 (20 °C)
Flash Pt: 191 °C
MDL Number: MFCD00026683
CAS Number: 32360-05-7
EINECS: 251-013-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cde19bfb96f7e2c53cdab70004edca070bb83b2d&vid=cde19bfb96f7e2c53cdab70004edca070bb83b2d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR