4-Methyl-1,2-cyclohexanedicarboxylic anhydride (mixture of cis- and trans-isomeres) ≥98.0%

Supplier: TCI America

M0567-25G M0567-500G
TCM0567-025GEA 33.18 CAD
TCM0567-025G TCM0567-500G
4-Methyl-1,2-cyclohexanedicarboxylic anhydride (mixture of cis- and trans-isomeres) ≥98.0%
4-Methyl-1,2-cyclohexanedicarboxylic anhydride (mixture of cis- and trans-isomeres)

CAS Number: 19438-60-9
MDL Number: MFCD00005927
Molecular Formula: C9H12O3
Molecular Weight: 168.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 164
Specific Gravity (20/20): 1.15

Formula: C₉H₁₂O₃
Density: 1.15 (20 °C)
Flash Pt: 164 °C
MDL Number: MFCD00005927
CAS Number: 19438-60-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa2ee5cf8f2813b88becb243d74c3e5d42fe6d93&vid=fa2ee5cf8f2813b88becb243d74c3e5d42fe6d93&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR