2-(Diethylamino)ethyl methacrylate ≥98.5% stabilized

Supplier: TCI America

M0562-500ML M0562-25ML
TCM0562-500MLEA 109.32 CAD
TCM0562-500ML TCM0562-025ML
2-(Diethylamino)ethyl methacrylate ≥98.5% stabilized
2-(Diethylamino)ethyl methacrylate

(stabilized with MEHQ)
CAS Number: 105-16-8
MDL Number: MFCD00038314
Molecular Formula: C10H19NO2
Molecular Weight: 185.27
Purity/Analysis Method: >98.5% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Flash Point (°C): 94
Specific Gravity (20/20): 0.92


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Methacrylic Acid 2-(Diethylamino)ethyl Ester

Formula: C₁₀H₁₉NO₂
Boiling Pt: 80 °C
Density: 0.92 (20 °C)
Flash Pt: 94 °C
MDL Number: MFCD00038314
CAS Number: 105-16-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e94db9166e4dd2d273025c22073f3bf17ba78d5&vid=3e94db9166e4dd2d273025c22073f3bf17ba78d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.5% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR