4-Methylphthalic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
M0560-25G
M0560-500G
TCM0560-025GEA
120.08
CAD
TCM0560-025G
TCM0560-500G
4-Methylphthalic acid ≥98.0% (by HPLC, titration analysis)
4-Methylphthalic acid
CAS Number: 4316-23-8
MDL Number: MFCD00041946
Molecular Formula: C9H8O4
Molecular Weight: 180.16
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 147
Flash Point (°C): 105
Formula:
C₉H₈O₄ Melting Pt: 147 °C Flash Pt: 105 °C |
MDL Number:
MFCD00041946 CAS Number: 4316-23-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...