Calcium methanesulfonate ≥98.0%

Supplier: TCI America

Synonyms: Methanesulphonic acid calcium salt, Methanesulfonic acid Calcium salt

M0549-25G M0549-500G
TCM0549-025GEA 221.78 CAD
TCM0549-025G TCM0549-500G
Calcium methanesulfonate ≥98.0%
Calcium methanesulfonate

CAS Number: 58131-47-8
MDL Number: MFCD00070543
Molecular Formula: CH4O3S
Molecular Weight: 230.26
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: Methanesulfonic Acid Calcium Salt

Formula: CH₄O₃S
MDL Number: MFCD00070543
CAS Number: 58131-47-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=615eff0cc244dca9144525d13add5adc2691504b&vid=615eff0cc244dca9144525d13add5adc2691504b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR